Order Entry
United States
ContactUsLinkComponent
Dextrose 5% in water for injection, sterile
Dextrose 5% in water for injection, sterile
Catalog # 68001-060
Supplier:  Baxter
CAS Number:  
Dextrose 5% in water for injection, sterile
Catalog # 68001-060
Supplier:  Baxter
Supplier Number:  2B0089
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bag, Viaflex®, multi pack
  • Size:
    100 ml
  • Cat. No.:
    68001-060
  • Supplier no.:
    2B0089

Specifications

About this item

 :
 :180.16 g/mol
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912