Order Entry
United States
ContactUsLinkComponent
 
Mannitol solution 5% in water, Osmitrol® USP for injection
Catalog # 68000-976
Supplier:  Baxter
CAS Number:  
Mannitol solution 5% in water, Osmitrol® USP for injection
Catalog # 68000-976
Supplier:  Baxter
Supplier Number:  2D5604X
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Bag, Viaflex®
  • Size:
    1000 ml
  • Cat. No.:
    68000-976
  • Supplier no.:
    2D5604X

Specifications

About this item

 :
 :182.17 g/mol
CAS Number:69-65-8
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00064287