Order Entry
United States
ContactUsLinkComponent
Sodium chloride solution 0.9% USP for irrigation
Sodium chloride solution 0.9% USP for irrigation
Catalog # 76313-488
Supplier:  B Braun
CAS Number:  
Sodium chloride solution 0.9% USP for irrigation
Catalog # 76313-488
Supplier:  B Braun
Supplier Number:  R8205
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Titan XL®
  • Cat. No.:
    76313-488

Specifications

About this item

 :
 :58.44 g/mol
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477