Metapenaeus Ensis Recombinant Met e 1 (from E. coli)
76778-238
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Metapenaeus ensis
- Size:250 µg
- Storage Conditions:–20…–80 °C
- Endotoxin Content:Please contact us for more information.
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:N
- Carrier-Free:Y
- Animal-Free:Y
- Protein/Peptide Name:Met e 1
- Purity:≥85% as determined by SDS-PAGE
- Molecular Weight:The recombinant Met e 1 protein predicts a molecular mass of 58.1 kDa
- Shipping Temperature:Ambient
- Cat. No.:76778-238