Human Recombinant SKP1 (from E. coli) Biotinylated
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human SKP1 (P63208) (Met1-Lys163) was expressed with a polyhistidine tag at the N-terminus and an AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Specifications
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20…–80 °C
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Gene ID:P63208
- Reconstitution Instructions:A hardcopy of datasheet with reconstitution instructions is sent along with the products. Please refer to it for detailed information.
- Endotoxin-free:N
- Carrier-Free:Y
- Animal-Free:Y
- Protein Synonyms:p19A|EMC19|OCP-II|TCEB1L|SKP1A|OCP2
- Protein/Peptide Name:SKP1
- Purity:>95% as determined by SDS-PAGE
- Molecular Weight:The recombinant human SKP1 consists of 190 amino acids and predicts a molecular mass of 22 kDa
- Sequence:Met1-Lys163
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together