ContactUsLinkComponent
miniPCR® Food Safety Lab: Mars Colony at Risk!
miniPCR® Food Safety Lab: Mars Colony at Risk!
Catalog # 470352-020
Supplier:  AMPLYUS LLC
undefined
miniPCR® Food Safety Lab: Mars Colony at Risk!
Catalog # 470352-020
Supplier:  AMPLYUS LLC
Supplier Number:  KT-1001-03
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    miniPCR® food safety lab
  • No. of lab groups:
    8
  • Cat. No.:
    470352-020
Specifications
About this item
Help astronauts control an outbreak of pathogenic bacteria in space food bound for Mars!
  • Techniques: Micropipetting, PCR, restriction digest, gel electrophoresis
  • Topics: Microbiology. infectious disease, molecular diagnostics, biotechnology
  • Time required: 120 minutes; can be split over 2 or 3 class periods
  • Level: General high school through advanced high school
Identify the source of pathogenic bacteria in space food bound for Mars! This applied biotechnology investigation addresses a foodborne illness outbreak using the same real-world methodologies that public health and surveillance authorities use here on Earth. Get hands-on experience with important molecular microbiology techniques and engage in a real-world problem with public health and economic impacts.
Reagents and consumables supplied by user:
- Agarose (electrophoresis grade)
- DNA stain (e.g., GelGreen™)
- Gel electrophoresis buffer (e.g., 1X TBE)
- Distilled or deionized H₂O (to dilute buffer concentrate – not included in Lab Companion Kit)
- Microtubes
- PCR tubes
Required equipment:
- Micropipettes and tips
- Thermal cycler
- Gel electrophoresis and visualization system
Storage notes:
- Reagents require freezer storage
- Reagents are stable for 12 months when stored in the freezer
Accessories information: The learning lab companion kit (catalog no. 470352-026) contains all the consumables (except water) needed for a class of 32 students. These reagents and consumables can also be sourced independently.
Delivery: The miniPCR Food Safety Lab kit contains reagents for 8 lab groups: 2X EZ PCR Master Mix, Load-Ready™; 3X Food Safety Lab Primer Mix; 4 DNA samples to investigate, including controls; Restriction enzyme XmnI; 100 bp DNA ladder, Load-Ready™. Note: Reagents are compatible with blueGel™ or other standard horizontal electrophoresis systems.