MULTISCREEN™ Dog 5-HT2A CHO-K1 HTS Membranes
Supplier: MULTISPAN INC
Certificates
About this item
Specifications
- Storage Conditions:LN₂ (Below –135 ℃)
Specifications
Related Information
MULTISCREEN™ Membranes Expressing GPCRs and Beyond
Application Notes
Multispan MULTISCREEN™ Products and Services
Product Literature
Frequently Bought Together