Order Entry
United States
ContactUsLinkComponent
miniPCR® P51™ Enzyme Lab: β-Gal Glow
miniPCR® P51™ Enzyme Lab: β-Gal Glow
Catalog # 470352-022
Supplier:  AMPLYUS LLC
undefined
miniPCR® P51™ Enzyme Lab: β-Gal Glow
Catalog # 470352-022
Supplier:  AMPLYUS LLC
Supplier Number:  KT-1900-02

Specifications

  • Description:
    miniPCR® P51™ enzyme lab: β-Gal Glow™
  • No. of lab groups:
    8
  • Cat. No.:
    470352-022

Specifications

About this item

Use an inquiry-based approach to investigate enzyme activity.

  • Techniques: Micropipetting, fluorescence detection
  • Topics: Enzymes, fluorescence, biotechnology
  • Time required: Can be completed in one 45-minute class
  • Level: General high school through college

Investigate enzyme activity and the conditions that affect it. Under optimal conditions, enzymes catalyze, or speed up the rate of, chemical reactions. In this lab, a substrate that is not fluorescent under basal conditions will fluoresce under blue light when sugar molecules are removed through hydrolysis by the enzyme beta-galactosidase. Observing how brightly the solution fluoresces over time allows students to see the reaction progress and quantify the effect of different variables on enzyme activity. This lab includes four inquiry-based investigations. Students explore the effects of pH, temperature, enzyme and substrate concentration, and competitive inhibition on enzyme reaction rate. This lab provides enough reagents for eight lab groups to each pursue at least two of these investigations.

Required equipment (one per lab group):
- Micropipettes and tips.
- P51™ Molecular fluorescence viewer or other blue light transilluminator (460 to 480 nm, e.g. blueBox™ or blueGel™).
- Suggested for temperature investigations: Thermal cycler or heat block that can accommodate PCR tubes (ideal setup) or water bath and floaties, ice (workable).

Storage notes:
- Reagents require refrigerator storage.
- Reagents are stable for 6 months when stored in the refrigerator.

Note: Reagents are compatible with P51™ molecular fluorescence viewer or other blue light transilluminator (460 to 480 nm, e.g. blueBox™ or blueGel™).

Delivery information: Includes substrate for enzymatic reaction; purified Beta-galactosidase enzyme solution; dilution buffers (2); inhibitor solutions (2); alkaline medium; fluorescent solution for standard curve.