ContactUsLinkComponent
HABITAT Sample Flask Rack for HABITAT research Bioreactors
HABITAT Sample Flask Rack for HABITAT research Bioreactors
Catalog # 76836-702
Supplier:  IKA Works
undefined
HABITAT Sample Flask Rack for HABITAT research Bioreactors
Catalog # 76836-702
Supplier:  IKA Works
Supplier Number:  20110862
Specifications
  • Description:
    HABITAT sample flask rack
  • Cat. No.:
    76836-702
Specifications
About this item
With the help of this stand, you can safely store your sample bottles.
There are four positions for 250 ml bottles. Due to a slightly inclined position, the sample bottle can be completely emptied without any problems.