ContactUsLinkComponent
ACCU-SCOPE® 3012 Upright Microscopes
ACCU-SCOPE® 3012 Upright Microscopes
Supplier:  ACCU-SCOPE INC SE
Certificates
undefined
ACCU-SCOPE® 3012 Upright Microscopes
Supplier:  ACCU-SCOPE INC SE
Specifications
  • Catalog No:
  • 470351-268
  • 470351-236
  • 470351-210
  • Type:
  • Binocular
  • Binocular
  • Binocular
  • Contrasting Technique:
  • Brightfield
  • Brightfield
  • Brightfield
  • Stage:
  • Rackless mechanical
  • Rackless mechanical
  • Rackless mechanical
  • Illumination:
  • LED
  • LED
  • LED