Engineered for classroom applications, the EXC-100 Series is built on a rugged, die-cast aluminum frame designed for years of use.
- Binocular or trinocular microscope
- Brightfield or Darkfield contrast method
- WF10×/18 mm eyepieces (one with pointer)
- 4×, 10×, 40×R and 100×R oil objectives
- Optional camera and/or camera with monitor
- Mechanical stage
- N.A. 1.25 Abbe condenser on spiral mount with iris
- Variable rechargeable LED illumination, with right angle power cord
- Cord wrap
Featuring ergonomic focusing controls, all metal focusing gears and a wide field of view, the EXC-100 Series delivers an exceptional balance of value and performance in an educational microscope. Choose between binocular (EXC-100) and trinocular (EXC-103) viewing models.
Ideally suited for the classroom environment, the EXC-100 Series provides clear images during observation. with front lens surfaces that are flush and easy to clean. EXC-100 microscopes come standard with Achromat 4×, 10×, 40× and 100x oil immersion DIN objectives.
The economical LED illuminator in the EXC-100 Series uses much less energy than traditional microscopes, plus it is rechargeable, so the microscope can be operated with or without a cord. The low positioned coaxial focusing mechanism provides complete ergonomic comfort to users during extended viewing periods. The coaxial focusing controls allow the user to rest their hand on the counter while focusing the microscope, allowing for maximum comfort.
- Catalog No:
- 470351-084
- 470351-252
- 470351-196
- 470351-186
- 470351-248
- 470351-126
- 470351-170
- 470351-192
- 470351-116
- Model:
- EXC-103-WIFI
- EXC-103-HDS
- EXC-103-HD
- EXC-103-EP
- EXC-103-EC50
- EXC-103-DF
- EXC-103
- EXC-100-DF
- EXC-100
- Contrasting Technique:
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield
- Brightfield and Darkfield
- Brightfield
- Brightfield and Darkfield
- Brightfield
- Stage:
- Mechanical
- Mechanical
- Mechanical
- Mechanical
- Mechanical
- Mechanical
- Mechanical
- Mechanical
- Mechanical
- Illumination:
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED
- LED