E.Z.N.A.® Plasmid DNA Mini Kit I, (Q-spin, capless)
Catalog # 76539-234
Supplier: Omega Bio-Tek
CAS Number:
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Plasmid
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:Mini plasmid kit
- Size:600 Preps
- Cat. No.:76539-234
Specifications
About this item
Isolate up to 25 µg plasmid DNA from 1 to 5 ml bacterial cultures using mini spin columns.
- Rapid - Purification of plasmid DNA in less than 30 minutes
- Safe - No Phenol/chloroform extractions
- Versatile - Spin and vacuum formats available
- High-quality - DNA is suitable for a variety of downstream applications
The E.Z.N.A.® Plasmid DNA Mini Kit I is designed to isolate up to 25 µg of high-quality plasmid DNA from 1 to 5ml bacterial cultures in less than 30 minutes. Plasmid DNA purification follows the alkaline-lysis method and is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, transformation, PCR and DNA sequencing.