ContactUsLinkComponent
VWR® PTFE Stopcock for VWR Separating Funnels
VWR® PTFE Stopcock for VWR Separating Funnels
  75838-904
VWR® PTFE Stopcock for VWR Separating Funnels
  75838-904
  • Description:
    Replacement stopcock
  • Material:
    PTFE
  • Cat. No.:
    75838-904
  • Bore size:
    12 mm
Replacement 12 mm stopcock for VWR® separating funnels.