Anti-Myc Tag Mouse Monoclonal Antibody [clone: mRV19]
Supplier: ReVacc Scientific
Certificates
About this item
This antibody has been tested to bind to Myc tagged protein.
Type: Primary
Antigen: Myc Tag
Clonality: Monoclonal
Clone: mRV19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Rat
Specifications
- Catalog No:
- 76729-642
- 76729-668
- Antigen Symbol:
- Myc Tag
- Myc Tag
- Antigen Name:
- Myc Tag
- Myc Tag
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- mRV19
- mRV19
- Reactivity:
- Rat
- Rat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2 kappa
- IgG2 kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Modification:
- Recombinant monoclonal antibody
- Recombinant monoclonal antibody
- Molecular Weight:
- 150 kDa
- 150 kDa
- Purification:
- Purified by Protein A chromatography
- Purified by Protein A chromatography
- Concentration:
- 1 mg/ml by refractometry
- 1 mg/ml by refractometry
- Storage Buffer:
- PBS, pH 7.2
- PBS, pH 7.2
- Storage Temperature:
- Store at 4 °C, −20 °C
- Store at 4 °C, −20 °C
- Shipping Temperature:
- Ice
- Ice
Specifications
Frequently Bought Together