ContactUsLinkComponent
Hanna Checker® pH Tester
undefined
Hanna Checker® pH Tester
  • Catalog No:
  • 470003-684
  • CA470003-684
  • pH Range:
  • 0.00 - 14.00
  • 0.00 - 14.00
  • pH Resolution:
  • 0.01
  • 0.01
  • pH Accuracy:
  • ±0.2
  • ±0.2
  • pH Calibration:
  • Manual, 2 points
  • Manual, 2 points
  • Battery Life:
  • approx. 3000 hours of continuous use
  • approx. 3000 hours of continuous use
  • Power supply:
  • 2 × 1.5 V alkaline batteries
  • 2 × 1.5 V alkaline batteries
  • Dimensions:
  • 66×50×25 mm (2.6×2.2×1")
  • 66×50×25 mm (2.6×2.2×1")
  • Weight:
  • 50 g (1.8 oz)
  • 50 g (1.8 oz)
Frequently Bought Together