Virus Recombinant SARS-CoV-2 Omicron BA.2.75 Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Supplier: ReVacc Scientific
Certificates
About this item
A recombinant form of the receptor binding domain (RBD) with amino acids 319 to 541) of the spike (S) glycoprotein gene from SARS-CoV-2 bearing mutations identified in variant of Omicron BA.2.75, was produced by insect cells, followed by purification.
- This product contains C-terminal 6 hexa-histidine tag
This product has been run on SDS-PAGE with reduced condition. We also evaluated its binding to ACE2-Fc, anti-His mAb and a negative mAb in ELISAs.
Specifications
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Virus
- Tag sequence:His Tag
- Storage Conditions:–80 °C
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:SARS-COV-2 spike receptor binding domain (His Tag) - Omicron BA.2.75 strain
- Protein/Peptide Name:SARS-COV-2 OMICRON BA.2.75 RBD
- Molecular Weight:~28 kDa
- Concentration:0.2 mg/ml
- Formulation:Frozen liquid, in PBS pH 7.5
- Nuclease-free:N
- Shipping Temperature:Dry Ice
- Tested Applications:ELISA, SDS-PAGE
Specifications
Frequently Bought Together