Order Entry
United States
ContactUsLinkComponent
Virus Recombinant SARS-CoV-2 Omicron Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Virus Recombinant SARS-CoV-2 Omicron Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Supplier:  ReVacc Scientific
Certificates
undefined
Virus Recombinant SARS-CoV-2 Omicron Spike Protein Receptor Binding Domain (RBD) (from Insect cell), His Tag
Supplier:  ReVacc Scientific

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Virus
  • Tag sequence:
    His Tag
  • Storage Conditions:
    –80 °C
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    SARS-COV-2 spike receptor binding domain (His Tag) - Omicron strain
  • Protein/Peptide Name:
    SARS-COV-2 OMICRON RBD (HIS TAG)
  • Molecular Weight:
    ~28 kDa
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Frozen liquid, in PBS pH 7.5
  • Nuclease-free:
    N
  • Shipping Temperature:
    Dry Ice
  • Tested Applications:
    ELISA, SDS-PAGE

Specifications

Frequently Bought Together