Order Entry
United States
ContactUsLinkComponent
Anti-Trastuzumab Mouse Monoclonal Antibody [clone: 15H2]
Anti-Trastuzumab Mouse Monoclonal Antibody [clone: 15H2]
Catalog # 76510-082
Supplier:  Genscript
Anti-Trastuzumab Mouse Monoclonal Antibody [clone: 15H2]
Catalog # 76510-082
Supplier:  Genscript
Supplier Number:  A02033-40
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Trastuzumab
  • Antigen Symbol:
    Trastuzumab
  • Clonality:
    Monoclonal
  • Clone:
    15H2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
  • Size:
    40 µg
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4, containing 0.02% sodium azide
  • Storage Temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Shipping Temperature:
    Ice
  • Immunogen:
    Trastuzumab
  • Purification:
    Protein A affinity column
  • Cat. No.:
    76510-082
  • Packaging:
    Vial

Specifications

About this item

The product is specific for trastuzumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A02032-40 anti-trastuzumab antibody (11C4), mAb, mouse.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.

Type: Primary
Antigen: Trastuzumab
Clonality: Monoclonal
Clone: 15H2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: