Order Entry
United States
ContactUsLinkComponent
Anti-Cetuximab Rabbit Monoclonal Antibody [clone: 90G3]
Anti-Cetuximab Rabbit Monoclonal Antibody [clone: 90G3]
Catalog # 76510-008
Supplier:  Genscript
Anti-Cetuximab Rabbit Monoclonal Antibody [clone: 90G3]
Catalog # 76510-008
Supplier:  Genscript
Supplier Number:  A01992-40

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Cetuximab
  • Antigen Symbol:
    Cetuximab
  • Clonality:
    Monoclonal
  • Clone:
    90G3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Size:
    40 µg
  • Cross Adsorption:
    No
  • Form:
    Lyophilized
  • Storage Buffer:
    PBS, pH 7.4, containing 0.02% sodium azide
  • Storage Temperature:
    The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
  • Concentration:
    Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
  • Shipping Temperature:
    Ice
  • Immunogen:
    Cetuximab
  • Purification:
    Protein A affinity column
  • Cat. No.:
    76510-008
  • Packaging:
    Vial

Specifications

About this item

The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01991-40, MonoRab™ anti-cetuximab antibody (69H10), mAb, rabbit.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Cetuximab
Clonality: Monoclonal
Clone: 90G3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: