Anti-Ipilimumab Mouse Monoclonal Antibody [clone: 26B6H7D9]
76197-314
- Antibody Type:Primary
- Antigen Name:Ipilimumab
- Antigen Symbol:Ipilimumab
- Clonality:Monoclonal
- Clone:26B6H7D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Size:40 µg
- Cross Adsorption:No
- Form:Lyophilized
- Storage Buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage Temperature:The lyophilized product remains stable up to 1 year at −20 °C from date of receipt. Upon reconstitution, it can be stored for 2 - 3 weeks at 2 - 8 °C or for up to 12 months at −20 °C or below. Avoid repeated freezing and thawing cycles
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to a final concentration of 0.5 mg/ml
- Shipping Temperature:Ice
- Immunogen:Ipilimumab
- Purification:Protein A affinity column
- Cat. No.:76197-314
- Packaging:Vial
The product is specific for Ipilimumab. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A01858-40, Anti- Ipilimumab Antibody (4H6E1D4), mAb, Mouse.
- Anti-Idiotype antibodies
- High specificit
- High affinity
- High stability
GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
Type: Primary
Antigen: Ipilimumab
Clonality: Monoclonal
Clone: 26B6H7D9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: