To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Secondary
- Antigen Name:IgG Fab
- Antigen Symbol:IgG Fab
- Clonality:Monoclonal
- Clone:12H3C4A6
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:200 µl
- Cross Adsorption:No
- Form:Liquid
- Storage Buffer:PBS (pH 7.4), 1% BSA, 0.01% thimerosal and 50% glycerol
- Storage Temperature:Store at −20 °C. This product is stable for 1 year upon receipt, when handled and stored as instructed
- Shipping Temperature:Ice
- Immunogen:Human IgG(H and L)
- Purification:Protein A affinity column
- Cat. No.:76197-306
- Packaging:Vial
Specifications
About this item
GenScript Mouse Anti-Human IgG Fab Antibody (12H3C4A6)[HRP], mAb reacts with the Fab portion of human IgG but not with the Fc portion of human IgG. The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken or goat immunoglobulins.
- High specificit
- High affinity
- High stability
At GenScript, secondary antibodies are purified from the serum of several hosts, including goat, donkey, mouse, rabbit, chicken and sheep. Secondary antibodies are available with specificity for whole Ig molecules or antibody fragments such as the Fc or Fab regions. The conjugated secondary antibodies can be used for various immunology analysis and protein assays.
Type: Secondary
Antigen: IgG Fab
Clonality: Monoclonal
Clone: 12H3C4A6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human