- Description:Multiparameter dissolved oxygen meter with DO electrode
- Operating Temperature Range:0...50 °C (32...122 °F)
- DO Range:0.00 - 45.00 ppm (mg/L), 0.0 - 300.0%, 0.0...50.0 °C (32.0...122.0 °F) (temperature limits will be reduced to actual probe/sensor limits)
- DO Resolution:0.01 ppm (mg/L); 0.1% saturation
- Conductivity Accuracy:±1% of reading (±0.05 μs/cm or 1 digit, whichever is greater)
- Conductivity Calibration:1 Cell factor calibration; 6 standards available: 84, 1413 μs/cm, 5.00, 12.88, 80.0, 111.8 ms/cm, 1 point offset: 0.00 μs/cm
- Conductivity Range:0.00 - 29.99 μs/cm, 3.00 - 29.99 μs/cm, 30.0 - 299.9 μs/cm, 300 - 2999 μs/cm, 3.00 - 29.99 ms/cm, 30.0 - 200.0 ms/cm, up to 500.0 ms/cm
- Conductivity Resolution:0.01 μs/cm, 00.1 μs/cm, 0.01 μs/cm, 0.01 mS/cm, 00.1 mS/cm
- DO Calibration:One or two points at 0% (HI 7040) and 100% (water saturated air)
- DO Accuracy:±1.5% of reading or ±1 digit
- TDS Accuracy:±1% of reading (±0.03 ppm or 1 digit, whichever is greater)
- TDS Calibration:Through EC calibration
- TDS Range:0.00 - 14.99 ppm (mg/L), 15.0 - 149.9 ppm (mg/L), 150. - 1499. ppm (mg/L), 1.50 - 14.99 g/L, 15.0 - 100.0 g/L, up to 400.0 g/L absolute TDS ‡ (with 0.80 factor)
- TDS Resolution:0.00 - 14.99 ppm (mg/L), 15.0 - 149.9 ppm (mg/L), 150. - 1499. ppm (mg/L), 1.50 - 14.99 g/L, 15.0 - 100.0 g/L, up to 400.0 g/L absolute TDS ‡ (with 0.80 factor)
- pH Accuracy:±0.01 pH, ±0.002 pH
- pH Calibration:Automatic, up to 3 points (5 points*) calibration, 5 standard (7 standard*) buffers available (1.68*, 4.01 or 3.00, 6.86, 7.01, 9.18, 10.01, 12.45*) and 2 custom buffers*
- pH Range:–2.000 to +16.000
- pH Resolution:0.001 pH, 0.01 pH
- Operating Humidity Range:RH max 95% non-condensing
- Cat. No.:470122-476
This laboratory benchtop dissolved oxygen meter delivers precision while offering simplicity and versatility in a sleek design. Use your multiparameter meter as a portable, benchtop meter, or mount it on the wall to save space.
- The large display and it's wide 150° viewing angle provide one of the easiest to read LCDs in the industry
- Setup and calibrate your meter in no time with as little as a 1 point calibration
- Great for use in business and educational laboratories worldwide
- Dissolved oxygen and conductivity electrodes purchased seperately to enable you to measure Conductivity, Total Dissolved Solids (TDS), and pH
- Your electronic pH meter will automatically recognize each digital electrode, providing sensor type, calibration data, and serial number Advanced pH electrode diagnostics ensure worry-free measurements
Wall Mount-Using the wall mount cradle (included), edge can be placed on a wall, leaving zero footprint on the benchtop space. The cradle has a built-in connector to power the meter and charge its batteries. edge's zero footprint is designed to save you valuable benchtop space.
Cradle and electrode holder - This meter is equipped with a benchtop cradle with an adjustable swivel electrode holder to charge and hold the meter securely in place at the optimum viewing angle.
Data logging - this digital pH meter allows you to store up to 1000 log records of data. Data sets include readings, GLP data, date and time.
DO: 0.00 to 45.00 ppm (mg/L), 0.0 to 300.0%, 0.0 to 50.0 °C (32.0 to 122.0 °F) (Temperature limits will be reduced to actual probe/sensor limits).
Conductivity: 0.00 to 29.99 μs/cm, 3.00 to 29.99 μS/cm, 30.0 to 299.9 μs/cm, 300 to 2999 μs/cm, 3.00 to 29.99 ms/cm, 30.0 to 200.0 ms/cm, up to 500.0 ms/cm.
TDS: 0.00 to 14.99 ppm (mg/L), 15.0 to 149.9 ppm (mg/L), 150 to 1499 ppm (mg/L), 1.50 to 14.99 g/L, 15.0 to 100.0 g/L, up to 400.0 g/L absolute TDS ‡ (with 0.80 factor).