- Description:pH Electrode Storage Solution
- Cat. No.:470122-066
- Supplier no.:HI70300M
This electrode storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes. Properly storing your pH electrode in a solution keeps the glass membrane well hydrated which maintains proper function and provides accurate readings.
- Special formulation to minimize bacterial growth on your electrode
- Designed for storing any pH or ORP electrode
- Air tight bottle with a tamper-proof seal Ensures quality and freshness of solution
This storage solution is a specially formulated electrode storage solution that can be used to store your pH electrode*. To ensure the best response time, the sensor tip and the reference junction of the pH electrode should be kept moist when not in use. Placing the pH electrode in a small glass filled with storage solution or replacing the solution in the protective cap is a n easy way to store the electrode. This storage solution should also be used to rehydrate the electrode after cleaning by soaking for at least one hour before taking measurements.