ContactUsLinkComponent
Hanna Instruments NaCl Solution
Hanna Instruments NaCl Solution
Catalog # 470122-054
Supplier:  HANNA INSTRUMENTS
CAS Number:  
Hanna Instruments NaCl Solution
Catalog # 470122-054
Supplier:  HANNA INSTRUMENTS
Supplier Number:  HI7081M
CAS Number:  
Specifications
  • Description:
    30 g/L NaCl Standard Solution (230 ml)
  • Cat. No.:
    470122-054
  • Supplier no.:
    HI7081M
Specifications
About this item
This premium quality 30 g/L NaCl standard solution for the calibration of salinity meters and sodium ISEs. Hanna calibration solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna's line of calibration solutions have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.
  • 29 g/L NaCl standard solution
  • Air tight bottle with tamper-proof seal to ensure quality
This premium quality 30 g/L NaCl calibration standard solution produced in accordance with ISO 3696/BS3978 standards using high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers. Reported values are traceable to NIST Standard Reference Materials (SRMs).