Order Entry
United States
Orders LinkContactUsLinkComponent
Hanna Instruments Zero Oxygen Solution Set (500 ml)

Hanna Instruments Zero Oxygen Solution Set (500 ml)

Zero oxygen solution set (500 ml)
Catalog # 470001-918
Supplier:   HANNA INSTRUMENTS
Supplier #:  HI7040L
Hanna Instruments Zero Oxygen Solution Set (500 ml)
Catalog # 470001-918
Supplier:  HANNA INSTRUMENTS
CAS Number:  
Hanna Instruments Zero Oxygen Solution Set (500 ml)
Hanna Instruments Zero Oxygen Solution Set (500 ml)

Zero oxygen solution set (500 ml)

CAS Number:  
Price
$26.09
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This is a high quality, bicomponent zero oxygen solution made from reagent grade chemicals. Hanna dissolved oxygen solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution.

  • Prepared with reagent grade chemicals
  • Hassle-free, easy to mix chemical component bottles
  • Air-tight seal to ensure the quality of the solution

This solution is a bicomponent zero oxygen solution designed for calibrating both polarographic and galvanic dissolved oxygen sensors. By simply immersing the dissolved oxygen probe in the solution users can quickly calibrate to 0% oxygen concentration, allowing for accurate follow-up measurements of dissolved oxygen in their sample. Hanna dissolved oxygen solutions are produced in our state-of-the-art manufacturing facility using reagent grade chemicals in a temperature controlled environment. 

Documents

Specifications

Specifications

  • Description:
    Zero oxygen solution set (500 ml)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.