- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:APLNR
- Description:Mouse APLNR ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Mouse APLNR ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse APLNR. No significant cross-reactivity or interference between APLNR and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:78.125 - 5000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:46.875 pg/ml
- Regulatory Status:RUO
- Cat. No.:76747-326
Mouse APLNR ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse APLNR in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse APLNR ELISA kit (A303566) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse APLNR in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for APLNR has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the APLNR present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-APLNR antibody, which binds the captured APLNR present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of APLNR captured in each well. The concentration of APLNR can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.