Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Cross Linked N-telopeptide of Type I Collagen/NTXI
- Description:Rat cross linked N-telopeptide of type I collagen/NTXI ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Rat Cross Linked N-telopeptide of Type I Collagen/NTXI ELISA kit exhibits high specificity and excellent specificity for the detection of rat Cross Linked N-telopeptide of Type I Collagen/NTXI. No significant cross-reactivity or interference between Cross Linked N-telopeptide of Type I Collagen/NTXI and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:3.125 - 200 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:1.875 ng/ml
- Regulatory Status:RUO
- Cat. No.:76741-976
Specifications
About this item
Rat Cross Linked N-telopeptide of Type I Collagen/NTXI ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Cross Linked N-telopeptide of Type I Collagen/NTXI in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 3.125 - 200 ng/ml
- Sensitivity: 1.875 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Rat Cross Linked N-telopeptide of Type I Collagen/NTXI ELISA kit (A79579) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Cross Linked N-telopeptide of Type I Collagen/NTXI in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Cross Linked N-telopeptide of Type I Collagen/NTXI has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cross Linked N-telopeptide of Type I Collagen/NTXI present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Cross Linked N-telopeptide of Type I Collagen/NTXI Antibody, which binds the captured Cross Linked N-telopeptide of Type I Collagen/NTXI present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Cross Linked N-telopeptide of Type I Collagen/NTXI captured in each well. The concentration of Cross Linked N-telopeptide of Type I Collagen/NTXI can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.