Order Entry
United States
Orders LinkContactUsLinkComponent
Mouse Complement Factor B ELISA Kit

Mouse Complement Factor B ELISA Kit

Mouse complement factor B ELISA kit, 96 tests
Catalog # 76735-136
Supplier:   Antibodies.com
Supplier #:  A77871-96
Mouse Complement Factor B ELISA Kit
Catalog # 76735-136
Supplier:  Antibodies.com
CAS Number:  
Mouse Complement Factor B ELISA Kit
Mouse Complement Factor B ELISA Kit

Mouse complement factor B ELISA kit, 96 tests

CAS Number:  
Price
$677.87
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Mouse Complement Factor B ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Complement Factor B in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 1.563 to 100 ng/ml
  • Sensitivity: 0.938 ng/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse Complement Factor B ELISA kit (A77871) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Complement Factor B in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Complement Factor B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Complement Factor B present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Complement Factor B Antibody, which binds the captured Complement Factor B present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Complement Factor B captured in each well. The concentration of Complement Factor B can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Mouse complement factor B ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    1.563 - 100 ng/ml
  • Sample Volume:
    100 μl
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76735-136
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.938 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Complement Factor B
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Mouse Complement Factor B ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse Complement Factor B. No significant cross-reactivity or interference between Complement Factor B and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.