Order Entry
United States
Orders LinkContactUsLinkComponent
Human KAT7/Hbo1/MYST2 ELISA Kit

Human KAT7/Hbo1/MYST2 ELISA Kit

Human KAT7 (Histone acetyltransferase KAT7) ELISA kit, 96 tests
Catalog # 76734-076
Supplier:   Antibodies.com
Supplier #:  A247377-96
Human KAT7/Hbo1/MYST2 ELISA Kit
Catalog # 76734-076
Supplier:  Antibodies.com
CAS Number:  
Human KAT7/Hbo1/MYST2 ELISA Kit
Human KAT7/Hbo1/MYST2 ELISA Kit

Human KAT7 (Histone acetyltransferase KAT7) ELISA kit, 96 tests

CAS Number:  
Price
$677.87
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human KAT7/Hbo1/MYST2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KAT7/Hbo1/MYST2 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human KAT7/Hbo1/MYST2 ELISA kit (A247377) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human KAT7/Hbo1/MYST2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for KAT7/Hbo1/MYST2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the KAT7/Hbo1/MYST2 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-KAT7/Hbo1/MYST2 Antibody, which binds the captured KAT7/Hbo1/MYST2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of KAT7/Hbo1/MYST2 captured in each well. The concentration of KAT7/Hbo1/MYST2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Human KAT7 (Histone acetyltransferase KAT7) ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    0.156 - 10 ng/ml
  • Sample Volume:
    100 μl
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76734-076
  • Regulatory Status:
    RUO
  • Sensitivity:
    0.094 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    KAT7
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Human KAT7/Hbo1/MYST2 ELISA Kit exhibits high specificity and excellent specificity for the detection of human KAT7/Hbo1/MYST2. No significant cross-reactivity or interference between KAT7/Hbo1/MYST2 and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.