Order Entry
United States
ContactUsLinkComponent
Rat Egr1 ELISA Kit
Rat Egr1 ELISA Kit
Catalog # 76745-182
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat Egr1 ELISA Kit
Catalog # 76745-182
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A79285-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Egr1
  • Description:
    Rat Egr1 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Rat Egr1 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Egr1. No significant cross-reactivity or interference between Egr1 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    6.25 - 400 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    3.75 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76745-182

Specifications

About this item

Rat Egr1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Egr1 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA kit
  • Detection Range: 6.25 to 400 pg/ml
  • Sensitivity: 3.75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Rat Egr1 ELISA kit (A79285) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat Egr1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Egr1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Egr1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Egr1 Antibody, which binds the captured Egr1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Egr1 captured in each well. The concentration of Egr1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.