- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Monkey
- Target protein:P-Selectin/CD62P
- Description:Monkey P-selectin/CD62P ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross Reactivity:Monkey P-Selectin/CD62P ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey P-Selectin/CD62P. No significant cross-reactivity or interference between P-Selectin/CD62P and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:31.25 - 2000 pg/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:18.75 pg/ml
- Regulatory Status:RUO
- Cat. No.:76743-342
Monkey P-Selectin/CD62P ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey P-Selectin/CD62P in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Monkey P-Selectin/CD62P ELISA kit (A303627) employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey P-Selectin/CD62P in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for P-Selectin/CD62P has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the P-Selectin/CD62P present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-P-Selectin/CD62P Antibody, which binds the captured P-Selectin/CD62P present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of P-Selectin/CD62P captured in each well. The concentration of P-Selectin/CD62P can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.