Order Entry
United States
ContactUsLinkComponent
Human Chorionic Gonadotropin IgG ELISA Kit
Human Chorionic Gonadotropin IgG ELISA Kit
Catalog # 76745-204
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Human Chorionic Gonadotropin IgG ELISA Kit
Catalog # 76745-204
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A247201-96
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Chorionic Gonadotropin IgG
  • Description:
    Human chorionic gonadotropin IgG ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Qualitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Storage Temperature:
    4 °C
  • Regulatory Status:
    RUO
  • Cat. No.:
    76745-204

Specifications

About this item

Human Chorionic Gonadotropin IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Chorionic Gonadotropin IgG in serum, plasma, tissue homogenates, and other biological fluids.