Order Entry
United States
ContactUsLinkComponent
Human XIAP ELISA Kit
Human XIAP ELISA Kit
Catalog # 76739-728
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human XIAP ELISA Kit
Catalog # 76739-728
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A247389-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    XIAP
  • Description:
    Human XIAP ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Human XIAP ELISA Kit exhibits high specificity and excellent specificity for the detection of human XIAP. No significant cross-reactivity or interference between XIAP and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76739-728

Specifications

About this item

Human XIAP ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human XIAP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human XIAP ELISA kit (A247389) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human XIAP in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for XIAP has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the XIAP present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-XIAP Antibody, which binds the captured XIAP present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of XIAP captured in each well. The concentration of XIAP can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.