Order Entry
United States
ContactUsLinkComponent
Rat Ovalbumin Specific IgG ELISA Kit
Rat Ovalbumin Specific IgG ELISA Kit
Catalog # 76737-528
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Rat Ovalbumin Specific IgG ELISA Kit
Catalog # 76737-528
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A303745-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Ovalbumin Specific IgG
  • Description:
    Rat ovalbumin specific IgG ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross Reactivity:
    Rat Ovalbumin Specific IgG ELISA Kit exhibits high specificity and excellent specificity for the detection of rat Ovalbumin Specific IgG. No significant cross-reactivity or interference between Ovalbumin Specific IgG and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.313 - 20 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.188 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76737-528

Specifications

About this item

Rat Ovalbumin Specific IgG ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat Ovalbumin Specific IgG in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.