Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Other
- Target protein:Angiotensin Converting Enzyme 1
- Description:Sheep angiotensin converting enzyme 1 ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Sheep Angiotensin Converting Enzyme 1 ELISA kit exhibits high specificity and excellent specificity for the detection of sheep Angiotensin Converting Enzyme 1. No significant cross-reactivity or interference between Angiotensin Converting Enzyme 1 and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.781 - 50 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.469 ng/ml
- Regulatory Status:RUO
- Cat. No.:76731-918
Specifications
About this item
Sheep Angiotensin Converting Enzyme 1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of sheep Angiotensin Converting Enzyme 1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.781 - 50 ng/ml
- Sensitivity: 0.469 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Sheep Angiotensin Converting Enzyme 1 ELISA kit (A76092) employs the sandwich enzyme immunoassay technique for the quantitative measurement of sheep Angiotensin Converting Enzyme 1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Angiotensin Converting Enzyme 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Angiotensin Converting Enzyme 1 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Angiotensin Converting Enzyme 1 Antibody, which binds the captured Angiotensin Converting Enzyme 1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Angiotensin Converting Enzyme 1 captured in each well. The concentration of Angiotensin Converting Enzyme 1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.