Order Entry
United States
ContactUsLinkComponent
Human C13orf33 ELISA Kit
Human C13orf33 ELISA Kit
Catalog # 76745-410
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human C13orf33 ELISA Kit
Catalog # 76745-410
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74354-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    C13orf33
  • Description:
    Human C13orf33 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human C13orf33 ELISA Kit exhibits high specificity and excellent specificity for the detection of human C13orf33. No significant cross-reactivity or interference between C13orf33 and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    78.125 - 5000 pg/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    46.875 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76745-410

Specifications

About this item

Human C13orf33 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human C13orf33 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 78.125 to 5000 pg/ml
  • Sensitivity: 46.875 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human C13orf33 ELISA Kit (A74354) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human C13orf33 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for C13orf33 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the C13orf33 present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-C13orf33 Antibody, which binds the captured C13orf33 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of C13orf33 captured in each well. The concentration of C13orf33 can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.