Order Entry
United States
ContactUsLinkComponent
Human Myosin Light Chain Kinase/MLCK ELISA Kit
Human Myosin Light Chain Kinase/MLCK ELISA Kit
Catalog # 76744-280
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human Myosin Light Chain Kinase/MLCK ELISA Kit
Catalog # 76744-280
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A74094-96
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Myosin Light Chain Kinase/MLCK
  • Description:
    Human myosin light chain kinase/MLCK ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    Human Myosin Light Chain Kinase/MLCK ELISA kit exhibits high specificity and excellent specificity for the detection of human Myosin Light Chain Kinase/MLCK. No significant cross-reactivity or interference between Myosin Light Chain Kinase/MLCK and analogues was observed.
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    0.156 - 10 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    100 μl
  • Sensitivity:
    0.094 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    76744-280

Specifications

About this item

Human Myosin Light Chain Kinase/MLCK ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Myosin Light Chain Kinase/MLCK in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-To-Use ELISA Kit
  • Detection Range: 0.156 - 10 ng/ml
  • Sensitivity: 0.094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Human Myosin Light Chain Kinase/MLCK ELISA kit (A74094) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Myosin Light Chain Kinase/MLCK in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for Myosin Light Chain Kinase/MLCK has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Myosin Light Chain Kinase/MLCK present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-Myosin Light Chain Kinase/MLCK Antibody, which binds the captured Myosin Light Chain Kinase/MLCK present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of Myosin Light Chain Kinase/MLCK captured in each well. The concentration of Myosin Light Chain Kinase/MLCK can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.