Order Entry
United States
Orders LinkContactUsLinkComponent
 

Human LCAT ELISA Kit

Human LCAT ELISA kit, 96 tests
Catalog # 76742-980
Supplier:   Antibodies.com
 :  A78380-96
Human LCAT ELISA Kit
Catalog # 76742-980
Supplier:  Antibodies.com
CAS Number:  
Human LCAT ELISA Kit
Human LCAT ELISA Kit

Human LCAT ELISA kit, 96 tests

CAS Number:  
Price
$677.87
Log in for availability
Each
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human LCAT ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human LCAT in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0.781 to 50 pg/ml
  • Sensitivity: 0.094 ng/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human LCAT ELISA kit (A78380) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human LCAT in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for LCAT has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the LCAT present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-LCAT Antibody, which binds the captured LCAT present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of LCAT captured in each well. The concentration of LCAT can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilized standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin Conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Human LCAT ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    0.781 - 50 pg/ml
  • Sample Volume:
    100 μl
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Assay Principle:
    Quantitative
  • Cat. No.:
    76742-980
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.094 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    LCAT
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Cross Reactivity:
    Human LCAT ELISA kit exhibits high specificity and excellent specificity for the detection of human LCAT. No significant cross-reactivity or interference between LCAT and analogues was observed.
  • Storage Temperature:
    4 °C

Specifications