Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:PI 3 Kinase Catalytic Subunit alpha/PIK3CA
- Description:Mouse PI 3 kinase catalytic subunit alpha/PIK3CA ELISA kit
- Size:96 tests
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:Mouse PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA kit exhibits high specificity and excellent specificity for the detection of mouse PI 3 Kinase Catalytic Subunit alpha/PIK3CA. No significant cross-reactivity or interference between PI 3 Kinase Catalytic Subunit alpha/PIK3CA and analogues was observed.
- Detection Method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:0.781 - 50 ng/ml
- Storage Temperature:4 °C
- Sample Volume:100 μl
- Sensitivity:0.469 ng/ml
- Regulatory Status:RUO
- Cat. No.:76732-260
Specifications
About this item
Mouse PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse PI 3 Kinase Catalytic Subunit alpha/PIK3CA in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-To-Use ELISA Kit
- Detection Range: 0.781 - 50 ng/ml
- Sensitivity: 0.469 ng/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Mouse PI 3 Kinase Catalytic Subunit alpha/PIK3CA ELISA kit (A75721) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse PI 3 Kinase Catalytic Subunit alpha/PIK3CA in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for PI 3 Kinase Catalytic Subunit alpha/PIK3CA has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the PI 3 Kinase Catalytic Subunit alpha/PIK3CA present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-PI 3 Kinase Catalytic Subunit alpha/PIK3CA Antibody, which binds the captured PI 3 Kinase Catalytic Subunit alpha/PIK3CA present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of PI 3 Kinase Catalytic Subunit alpha/PIK3CA captured in each well. The concentration of PI 3 Kinase Catalytic Subunit alpha/PIK3CA can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.