nUVaClean™ UV Pipette Sterilizer
Supplier: MTC BIO, INC.
Certificates
About this item
MTC’s nUVaClean™ is the first pipette rack that not only organizes and protects six pipettes, but also destroys greater than 99% of unwanted contamination.
- UV-C lamp and reflector system destroys 99% of DNA
- Eliminates cross-contamination
- 360° UV exposure
- Fits all major brands
- Compact 6.5'' (17 cm) footprint
The new nUVaClean™ uses a germicidal UV-C lamp. A high-efficiency reflector/concentrator completely bathes pipette shafts in 254 nm UV light. This exposure destroys unwanted microbiological contaminants and prevents cross-contamination in PCR and other sensitive procedures. Just place single-channel pipettes of any range or brand in the nUVaClean™ and press the 'start' button to begin the auto-decon cycle. After 28 minutes the nUVaClean will power down automatically, delivering up to six pipettes that are safe, clean and ready for use.
Frequently Bought Together