Human Recombinant CSNK2A1 (from E. coli), His
: Biortus
: Frozen liquid on dry ice.
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Gene ID:UniProt P68400
- Protein/Peptide Name:CSNK2A1
- Purity:90%
- Sequence:V5-S337
- Formulation:50 mM Tris-HCl (pH7.5), 200 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together