Human Recombinant CSK (from Insect cells), GST
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P41240
- Protein Synonyms:Tyrosine-protein kinase CSK
- Protein/Peptide Name:CSK
- Purity:90%
- Sequence:M1-L450
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 1 mM DTT, 5% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together