Human Recombinant CAMK2A (from Insect cells), GST
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9UQM7
- Protein/Peptide Name:CAMK2A
- Purity:90%
- Sequence:A2-H478
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 20% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Liquid. It is shipped out with dry ice
Specifications
Frequently Bought Together