Human Recombinant BRSK1 (from Insect cells), GST
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q8TDC3
- Protein/Peptide Name:BRSK1
- Purity:90%
- Sequence:S2-P778
- Formulation:50 mM Tris-HCl (pH 7.5), 150 mM NaCl, 5% glycerol, 1 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together