Human Recombinant AKT3 (from Insect cells), GST
: Biortus
: Frozen liquid on dry ice.
- Conjugation:GST
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9Y243
- Protein/Peptide Name:AKT3
- Purity:90%
- Sequence:S2-E479
- Formulation:50 mM Tris-HCl (pH 7.5), 500 mM NaCl, 5% glycerol, 5 mM DTT, 0.1 M Trehalose
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together