Human Recombinant TRPV3 (from Mammalian cells), Flag
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Flag
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:Human
- Gene ID:UniProt Q8NET8
- Protein/Peptide Name:TRPV3
- Purity:95%
- Sequence:K2-V790 end
- Formulation:50 mM Tris pH 8.0, 150 mM NaCl, 0.025% (w/v) DDM, 0.0025% CHS, 0.5 mM TCEP, 0.1 mg/ml POPC: POPE: POPG (3:1:1)
- Shipping Temperature:Liquid. It is shipped out with dry ice
Specifications
Frequently Bought Together