Human Recombinant CXCR4 (from Insect cells), Flag/His
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P61073
- Protein Synonyms:Membrane protein
- Protein/Peptide Name:CXCR4
- Purity:95%
- Sequence:E2-S229, K230-V320, L125W
- Formulation:25 mM HEPES pH 7.5, 150 mM NaCl, 0.025% (w/v) DDM, 0.005% (w/v) CHS, 1 μM IT1t
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together