Human Recombinant ADORA2A (from Insect cells), Flag/His
Supplier: Biortus
Certificates
About this item
Delivery information: Frozen liquid on dry ice.
Specifications
- Conjugation:Flag/His
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt P29274
- Protein Synonyms:Membrane protein
- Protein/Peptide Name:ADORA2A
- Purity:95%
- Sequence:P2-L208, E219-A316, S91K
- Formulation:20 mM HEPES pH 7.5, 500 mM NaCl, 20 μM theophylline, 10% glycerol, 0.05% (w/v) LMNG, 0.005% (w/v) CHS
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together