Human Recombinant ZFAND5 (from Insect cells), Unconjugated
: Biortus
: Frozen liquid on dry ice.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt O76080
- Protein Synonyms:Zinc finger protein 216
- Protein/Peptide Name:ZFAND5
- Purity:85%
- Sequence:A2-I213
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Frequently Bought Together