Human Recombinant ZFAND3 (from Insect cells), Unconjugated
Supplier: Biortus
Certificates
About this item
Delivery: Frozen liquid on dry ice.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Insect cells
- Species:Human
- Gene ID:UniProt Q9H8U3
- Protein/Peptide Name:ZFAND3
- Purity:85%
- Sequence:G2-S227
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 20% glycerol
- Shipping Temperature:Frozen liquid with dry ice
Specifications
Frequently Bought Together